Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003664.2 | NP_001003664.1 | 5 | 638 |
Gene Symbol: Try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038996.2 | NP_001034085.1 | 5 | 722 | ||
GenBank mRNA | AK133516.1 | - | 5 | 649 | ||
BC138698.1 | - | 5 | 722 | 8861720 | ||
BC146017.1 | - | 5 | 722 | 40131320 |