Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 9, 10 | 1415 | ||
NM_001322855.1 | NP_001309784.1 | 10, 11 | 1557 | |||
NM_001322856.1 | NP_001309785.1 | 10, 11 | 1514 | |||
NM_001322857.1 | NP_001309786.1 | 9, 10 | 1335 | |||
NM_001322858.1 | NP_001309787.1 | 10, 11 | 1432 | |||
NM_001322859.1 | NP_001309788.1 | 7, 8 | 1041 | |||
NM_001322860.1 | NP_001309789.1 | 6, 7 | 981 | |||
NM_001322861.1 | NP_001309790.1 | 6, 7 | 691 | |||
NM_001322862.1 | NP_001309791.1 | 10, 11 | 1392 | |||
NM_001322863.1 | NP_001309792.1 | 9, 10 | 1364 | |||
NM_001322864.1 | NP_001309793.1 | 8, 9 | 932 | |||
NM_001322866.1 | NP_001309795.1 | 9, 10 | 1255 | |||
NM_020448.4 | NP_065181.1 | 9, 10 | 1277 | |||
NR_136506.1 | 8, 9 | 1185 | ||||
NR_136508.1 | 8, 9 | 1146 | ||||
XM_011541808.2 | XP_011540110.1 | 8, 9 | 1192 | |||
XM_017001868.1 | XP_016857357.1 | 8, 9 | 927 | |||
XM_017001869.1 | XP_016857358.1 | 9, 10 | 1262 | |||
XM_017001870.1 | XP_016857359.1 | 10, 11 | 1527 | |||
GenBank mRNA | AK312433.1 | - | 9, 10 | 1231 | ||
BC063583.1 | - | 9, 10 | 1207 | 4386115 | ||
BX538110.1 | - | 3, 4 | 305 | |||
BX640883.1 | - | 10, 11 | 1339 | |||
U90918.1 | - | 9, 10 | 1168 |