Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP10D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020453.3 | NP_065186.3 | 8 | 1209 | ||
XM_005248119.4 | XP_005248176.1 | 8 | 1340 | |||
XM_005248120.4 | XP_005248177.1 | 8 | 1327 | |||
XM_011513722.2 | XP_011512024.1 | 7 | 1145 | |||
XM_017008472.1 | XP_016863961.1 | 8 | 1320 | |||
GenBank mRNA | AJ441078.1 | - | 8 | 1209 | ||
BC057839.1 | - | 8 | 1303 | |||
BC131535.1 | - | 8 | 1187 | 40082621 |