Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009950.1 | NP_001009950.1 | 11 | 2283 | ||
GenBank mRNA | BC089013.1 | - | 11 | 2283 | 30605563 |