Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127482.2 | NP_001120954.1 | 4 | 1467 | ||
NM_020456.3 | NP_065189.1 | 5 | 1584 | |||
NR_023351.2 | 3 | 1300 | ||||
GenBank mRNA | AF052154.1 | - | 5 | 1383 | ||
AF055016.1 | - | 5 | 1383 | |||
AF334405.1 | - | 5 | 1422 | |||
BC022519.1 | - | 4 | 1273 | 4792366 |