Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020525.4 | NP_065386.1 | 4, 5 | 508 | ||
GenBank mRNA | AF279437.1 | - | 4, 5 | 512 | ||
AJ277247.1 | - | 5, 6 | 526 | |||
AY358890.1 | - | 4, 5 | 512 | |||
BC066263.1 | - | 4, 5 | 505 | 6971854 | ||
BC066264.1 | - | 4, 5 | 504 | 6971855 | ||
BC066265.1 | - | 4, 5 | 505 | 6971857 | ||
BC067509.1 | - | 4, 5 | 505 | 6971858 | ||
BC067510.1 | - | 4, 5 | 505 | 6971859 | ||
BC069112.1 | - | 5, 6 | 526 | 7216929 | ||
BC069308.1 | - | 4, 5 | 505 | 7262127 | ||
BC070261.1 | - | 5, 6 | 523 | 30416027 | ||
CD559617.2 | - | 4, 5 | 535 |