Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006342.2 | NP_006333.1 | 2, 3 | 325 | ||
XM_005247929.2 | XP_005247986.1 | 2, 3 | 323 | |||
XM_005247930.2 | XP_005247987.1 | 2, 3 | 432 | |||
XM_011513386.2 | XP_011511688.1 | 2, 3 | 323 | |||
XM_017007653.1 | XP_016863142.1 | 2, 3 | 377 | |||
GenBank mRNA | AF093543.1 | - | 2, 3 | 278 | ||
AF479620.1 | - | 2, 3 | 270 | |||
AJ243997.1 | - | 2, 3 | 270 | |||
AK225631.1 | - | 2, 3 | 293 | |||
AK302463.1 | - | 2, 3 | 296 | |||
AK304579.1 | - | 1, 2 | 54 | |||
AK310660.1 | - | 1, 2 | 48 | |||
BC106071.1 | - | 2, 3 | 286 | 6148176 | ||
BC111771.1 | - | 2, 3 | 238 | 40034847 | ||
DB098941.1 | - | 2, 3 | 323 |