Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013370.2 | NP_001013388.2 | 10 | 1622 | ||
NM_001162908.1 | NP_001156380.1 | 10 | 2192 | |||
GenBank mRNA | AK159531.1 | - | 10 | 1568 | ||
AK160083.1 | - | 10 | 1583 | |||
BC055753.1 | - | 10 | 1612 | 6414521 | ||
BC100403.1 | - | 10 | 2192 | 30432354 |