Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013375.1 | NP_001013393.1 | 7 | 1032 | ||
GenBank mRNA | AK032265.1 | - | 7 | 976 | ||
AK078033.1 | - | 2 | 168 | |||
AK154637.1 | - | 7 | 972 | |||
AK167248.1 | - | 7 | 1031 | |||
BC119621.1 | - | 7 | 1005 | 40062238 | ||
BC119622.1 | - | 7 | 1005 | 40062245 |