Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017429.2 | NP_001017429.1 | 3 | 423 | ||
GenBank mRNA | BG792513.1 | - | 3 | 355 |