Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025353.2 | NP_001020524.1 | 2 | 157 | ||
GenBank mRNA | AK136644.1 | - | 2 | 158 | ||
AY621337.1 | - | 1 | 13 | |||
BC139164.1 | - | 1 | 106 | 8862186 | ||
DQ012051.1 | - | 1 | 13 |