Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031622.2 | NP_001026793.2 | 3 | 564 | ||
GenBank mRNA | AK145705.1 | - | 3 | 577 | ||
BC099967.1 | - | 3 | 564 | 30027969 |
Gene Symbol: Gm16513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200055.1 | NP_001186984.1 | 575 |
Gene Symbol: Gm3106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC061196.1 | - | 669 |
Gene Symbol: Gm3139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243937.1 | NP_001230866.1 | 3 | 595 | ||
NM_001243938.1 | NP_001230867.1 | 3 | 555 | |||
GenBank mRNA | CN708470.1 | - | 3 | 580 | ||
CN715963.1 | - | 3 | 555 |