Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033271.4 | NP_001028443.1 | 7 | 1227 | ||
GenBank mRNA | AK144251.1 | - | 7 | 1227 | ||
AK152640.1 | - | 7 | 1212 | |||
AK152658.1 | - | 7 | 1212 | |||
AK159641.1 | - | 7 | 1231 | |||
AK169026.1 | - | 3 | 2854 | |||
AK206063.1 | - | 1 | 23 | |||
BC027355.1 | - | 7 | 1212 | |||
BC039161.1 | - | 6 | 1064 | |||
BC049945.1 | - | 6 | 1084 | |||
BC058171.1 | - | 7 | 1176 | |||
BC106163.1 | - | 7 | 1206 | 4913757 |