Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033367.3 | NP_001028539.1 | 5 | 2516 | ||
GenBank mRNA | AK035300.1 | - | 2 | 1917 | ||
AK143864.1 | - | 5 | 2517 | |||
BC144861.1 | - | 3 | 2287 | |||
BC144862.1 | - | 3 | 2273 | |||
BC145617.1 | - | 4 | 2310 |