Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033367.3 | NP_001028539.1 | 9 | 3119 | ||
GenBank mRNA | AK035300.1 | - | 6 | 2520 | ||
AK143864.1 | - | 9 | 3120 | |||
AK171624.1 | - | 9 | 3114 | |||
BC144861.1 | - | 8 | 2968 | |||
BC144862.1 | - | 7 | 2876 | |||
BC145617.1 | - | 8 | 2913 |