Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286094.1 | NP_001273023.1 | 4 | 1917 | ||
NM_001286095.1 | NP_001273024.1 | 5 | 1566 | |||
NM_020644.2 | NP_065695.1 | 5 | 2009 | |||
GenBank mRNA | AK074677.1 | - | 5 | 1627 | ||
AY359069.1 | - | 5 | 1652 | |||
BC015884.1 | - | 4 | 1544 | 3909465 | ||
BC040124.1 | - | 5 | 1690 | 6053804 |