Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smagp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033872.2 | NP_001029044.1 | 4 | 597 | ||
NM_001285413.1 | NP_001272342.1 | 3 | 580 | |||
NM_001285414.1 | NP_001272343.1 | 3 | 533 | |||
NM_174992.4 | NP_778157.2 | 4 | 787 | |||
GenBank mRNA | BC004728.1 | - | 4 | 588 | 3582955 | |
BE132795.1 | - | 4 | 453 | |||
CK129185.1 | - | 4 | 491 |