Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033874.2 | NP_001029046.2 | 3 | 215 | ||
GenBank mRNA | BC100481.1 | - | 3 | 212 | 30917849 | |
BC109352.2 | - | 3 | 202 | 40057006 | ||
BY100469.1 | - | 3 | 216 |