Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145863.1 | NP_001139335.1 | 4 | 1461 |
Gene Symbol: Gm14124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142410.1 | NP_001135882.1 | 4 | 2129 |
Gene Symbol: Zfp345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034900.3 | NP_001030072.2 | 4 | 1627 | ||
GenBank mRNA | AK144819.1 | - | 4 | 1628 | ||
BC150850.1 | - | 4 | 1616 | 9087761 | ||
BC150852.1 | - | 4 | 1616 | 9087763 |