Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037221.2 | NP_001032298.1 | 8 | 2169 | ||
NM_001163433.1 | NP_001156905.1 | 7 | 1649 | |||
NM_028966.3 | NP_083242.1 | 7 | 1905 | |||
GenBank mRNA | AK017081.1 | - | 3 | 483 | ||
AK034323.1 | - | 6 | 1499 | |||
AK133470.1 | - | 7 | 1884 | |||
AK134243.1 | - | 8 | 2167 | |||
AK161277.1 | - | 7 | 1643 | |||
AK163751.1 | - | 6 | 1926 | |||
BC049740.1 | - | 3 | 455 | |||
BC145827.1 | - | 7 | 1771 | 40131130 | ||
BC145829.1 | - | 7 | 1770 | 40131132 | ||
DQ278487.1 | - | 7 | 1682 |