Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037955.4 | NP_001033044.1 | 4, 5 | 261 | ||
NM_134068.3 | NP_598829.1 | 4, 5 | 261 | |||
GenBank mRNA | AF237619.1 | - | 4, 5 | 221 | ||
AK148708.1 | - | 4, 5 | 229 | |||
AK149363.1 | - | 4, 5 | 220 | |||
BC057944.1 | - | 4, 5 | 231 | |||
BC080718.1 | - | 4, 5 | 211 | |||
BC108362.1 | - | 4, 5 | 211 | 4917645 | ||
CX236611.1 | - | 4, 5 | 261 |