Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA068307.1 | - | 2 | 200 | ||
BC028874.1 | - | 2 | 154 | 4194069 | ||
CJ183794.1 | - | 2 | 255 | |||
EF437365.1 | - | 2 | 161 | |||
X61431.1 | - | 2 | 173 |