Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008545.2 | NP_032571.2 | 1 | 1946 | ||
GenBank mRNA | AF311315.3 | - | 1 | 1214 | ||
AF311316.2 | - | 1 | 1214 | |||
AK019658.1 | - | 1 | 1946 | |||
AY099462.1 | - | 1 | 1214 | |||
AY099463.1 | - | 1 | 1214 | |||
AY099464.1 | - | 1 | 1214 |