Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbmxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252089.1 | NP_001239018.1 | 2 | 369 | ||
NM_009033.2 | NP_033059.2 | 2 | 415 | |||
GenBank mRNA | AF031568.1 | - | 2 | 281 | ||
AK041927.1 | - | 2 | 306 | |||
AK088213.1 | - | 2 | 302 | |||
AK149658.1 | - | 2 | 388 | |||
AK149764.1 | - | 2 | 388 | |||
AK214325.1 | - | 1 | 163 | |||
AK218950.1 | - | 1 | 165 | |||
BC011441.1 | - | 2 | 320 | 3153831 | ||
BC089350.1 | - | 2 | 329 | 30359767 | ||
BY262810.1 | - | 2 | 369 |