Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167578.1 | NP_001161050.1 | 12 | 2056 | ||
GenBank mRNA | AK076614.1 | - | 12 | 2002 | ||
AK132653.1 | - | 13 | 2152 |
Gene Symbol: Tcp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011553.4 | NP_035683.2 | 12 | 1956 | ||
GenBank mRNA | AK161455.1 | - | 12 | 1927 | ||
X58169.1 | - | 12 | 1821 |
Gene Symbol: Tcp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009341.2 | NP_033367.2 | 12 | 1962 | ||
GenBank mRNA | M22602.1 | - | 1 | 495 |