Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009450.2 | NP_033476.1 | 2, 3 | 247 | ||
GenBank mRNA | AK134545.1 | - | 2, 3 | 246 | ||
BC055441.1 | - | 2, 3 | 237 | 4016867 | ||
CJ148662.1 | - | 2, 3 | 280 |