Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009999.4 | NP_034129.1 | 4 | 635 | ||
GenBank mRNA | AF128849.1 | - | 4 | 617 | ||
AF129405.1 | - | 3 | 431 | |||
AI304065.1 | - | 4 | 587 | |||
AK028103.1 | - | 4 | 635 | |||
BC060973.1 | - | 4 | 629 | 30301385 | ||
M21856.1 | - | 4 | 617 | |||
X99715.1 | - | 4 | 613 |