Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010190.1 | NP_034320.1 | 2 | 128 | ||
GenBank mRNA | AF063217.1 | - | 2 | 101 | ||
AK010913.1 | - | 1 | 126 | |||
AK151676.1 | - | 2 | 136 | |||
BC107220.1 | - | 2 | 128 | 40051808 | ||
BC107221.1 | - | 2 | 128 | 40051810 |