Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020672.2 | NP_065723.1 | 4 | 680 | ||
XM_005245362.1 | XP_005245419.1 | 4 | 676 | |||
XM_017001875.1 | XP_016857364.1 | 5 | 768 | |||
GenBank mRNA | AY007220.1 | - | 4 | 662 |