Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010505.2 | NP_034635.2 | 1 | 324 | ||
GenBank mRNA | BC120910.1 | - | 1 | 324 | 40091152 | |
BC120911.1 | - | 1 | 324 | 40091153 |
Gene Symbol: Ifna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC117774.1 | - | 324 | 40091177 |