Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111274.1 | NP_001104744.1 | 3, 4 | 606 | ||
NM_010512.4 | NP_034642.2 | 3, 4 | 688 | |||
NM_184052.3 | NP_908941.1 | 3, 4 | 606 | |||
GenBank mRNA | AK038119.1 | - | 3, 4 | 1689 | ||
AY878193.1 | - | 3, 4 | 486 | |||
BC012409.1 | - | 2, 3 | 343 | 4194295 | ||
CT010364.1 | - | 2, 3 | 316 | |||
X04482.1 | - | 3, 4 | 388 |