Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010667.2 | NP_034797.1 | 4 | 792 | ||
GenBank mRNA | AK161149.1 | - | 4 | 794 | ||
BC139222.1 | - | 4 | 770 | 8862244 | ||
X99143.1 | - | 4 | 629 |
Gene Symbol: 5430421N21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201323.1 | NP_001188252.1 | 4 | 776 | ||
GenBank mRNA | AK133718.1 | - | 4 | 777 | ||
M92088.1 | - | 4 | 761 |
Gene Symbol: Krt81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166157.1 | NP_001159629.1 | 4 | 787 | ||
GenBank mRNA | AF312018.1 | - | 4 | 448 |