Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010691.5 | NP_034821.2 | 2 | 453 | ||
GenBank mRNA | AB108497.1 | - | 2 | 453 | ||
AK144562.1 | - | 2 | 298 | |||
BC119177.1 | - | 2 | 542 | 8733926 | ||
BC120586.1 | - | 2 | 542 | 40129509 | ||
X90829.1 | - | 1 | 1586 |