Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011963.2 | NP_036093.2 | 2 | 450 | ||
GenBank mRNA | AF128236.1 | - | 2 | 318 | ||
BC138857.1 | - | 2 | 384 | 8861879 | ||
BY713630.1 | - | 2 | 452 | |||
CN704107.1 | - | 2 | 436 | |||
M83346.1 | - | 2 | 318 |