Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146153.1 | NP_001139625.1 | 6 | 637 | ||
NM_011984.2 | NP_036114.2 | 6 | 637 | |||
GenBank mRNA | AF093261.1 | - | 4 | 464 | ||
AK179223.1 | - | 4 | 324 | |||
BC005773.1 | - | 6 | 586 | 3602414 | ||
BU054773.1 | - | 4 | 341 | |||
BU707796.1 | - | 1 | 65 | |||
CN539202.1 | - | 4 | 335 |