Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271496.1 | NP_001258425.1 | 2 | 726 | ||
NM_013490.4 | NP_038518.2 | 2 | 726 | |||
GenBank mRNA | AB011002.1 | - | 2 | 585 | ||
AK005057.1 | - | 2 | 727 | |||
AK014174.1 | - | 2 | 689 | |||
AK052056.1 | - | 2 | 668 | |||
AK053818.1 | - | 2 | 668 | |||
AK166465.1 | - | 2 | 667 | |||
AK166641.1 | - | 2 | 665 | |||
BC056758.1 | - | 2 | 613 | |||
BC060218.1 | - | 2 | 689 |