Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015740.3 | NP_056555.2 | 3, 4 | 352 | ||
GenBank mRNA | AK003035.1 | - | 3, 4 | 353 | ||
BC034662.1 | - | 3, 4 | 296 | 5359972 |