Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018860.4 | NP_061348.1 | 4 | 426 | ||
GenBank mRNA | AK007874.1 | - | 4 | 374 | ||
AK009548.1 | - | 4 | 373 | |||
AK010246.1 | - | 4 | 373 | |||
AK010482.1 | - | 4 | 373 | |||
AK010509.1 | - | 4 | 373 | |||
AK010564.1 | - | 4 | 371 | |||
AK010583.1 | - | 4 | 373 | |||
AK010585.1 | - | 4 | 371 | |||
AK012450.1 | - | 4 | 376 | |||
AK012476.1 | - | 4 | 372 | |||
AK012487.1 | - | 4 | 371 | |||
AK019049.1 | - | 4 | 373 | |||
AK019178.1 | - | 4 | 368 | |||
AK150502.1 | - | 4 | 373 | |||
AK151566.1 | - | 4 | 372 | |||
AK198849.1 | - | 1 | 141 | |||
AV478133.1 | - | 4 | 359 | |||
BC081450.1 | - | 3 | 442 | |||
BC083066.1 | - | 4 | 360 | |||
BC089572.1 | - | 4 | 370 | |||
BC089573.1 | - | 4 | 372 | |||
BC092010.1 | - | 4 | 361 | |||
BC099380.1 | - | 3 | 440 |