Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286168.1 | NP_001273097.1 | 7 | 874 | ||
NM_019733.3 | NP_062707.1 | 8 | 1008 | |||
GenBank mRNA | AF148511.1 | - | 8 | 957 | ||
AK079238.1 | - | 7 | 818 | |||
BC030397.1 | - | 8 | 969 | 4225078 | ||
BE625909.1 | - | 1 | 82 | |||
BY708385.1 | - | 5 | 502 |