Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002892.1 | - | 15 | 1506 | ||
AK009717.1 | - | 15 | 1505 | |||
AK076513.1 | - | 16 | 1596 | |||
AK088082.1 | - | 15 | 1502 | |||
BC021763.1 | - | 15 | 1532 | 4458575 |