Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207156.2 | NP_997039.2 | 1 | 284 |
Gene Symbol: Olfr749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020288.2 | NP_064684.2 | 2 | 350 |
Set Membership: |
G-protein coupled receptor |