Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162858.1 | NP_001156330.1 | 4 | 826 | ||
NM_021717.2 | NP_068363.2 | 4 | 826 | |||
GenBank mRNA | AJ278170.1 | - | 4 | 849 | ||
BC094230.1 | - | 3 | 1949 | 30535040 | ||
BC115523.1 | - | 4 | 576 | 40101227 |