Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022033.4 | NP_071316.1 | 1 | 1460 | ||
GenBank mRNA | AB022180.2 | - | 1 | 1398 | ||
AB045847.1 | - | 1 | 478 | |||
AK077075.1 | - | 1 | 1413 | |||
BC137887.1 | - | 1 | 1460 | 8860906 | ||
BC137888.1 | - | 1 | 1460 | 8860907 |