Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020726.4 | NP_065777.1 | 6 | 974 | ||
XM_005248559.2 | XP_005248616.1 | 6 | 965 | |||
XM_006714661.2 | XP_006714724.1 | 7 | 1029 | |||
XM_017009673.1 | XP_016865162.1 | 7 | 1029 | |||
GenBank mRNA | AB033052.2 | - | 6 | 796 | ||
AJ300837.1 | - | 6 | 921 | |||
AK096416.1 | - | 6 | 919 | |||
AK292803.1 | - | 6 | 915 | |||
BC001644.2 | - | 6 | 912 | 2958628 | ||
BC004985.1 | - | 6 | 912 | 2958628 |