Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bend5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026279.3 | NP_080555.1 | 6 | 1414 | ||
GenBank mRNA | AK075885.1 | - | 6 | 1414 | ||
AK156708.1 | - | 6 | 1316 | |||
AK167425.1 | - | 6 | 1234 | |||
BC037633.1 | - | 6 | 1247 | |||
BC051512.1 | - | 4 | 835 | 1383740 |