Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026445.2 | NP_080721.1 | 9 | 1315 | ||
GenBank mRNA | AK076022.1 | - | 9 | 1321 | ||
AK138300.1 | - | 9 | 1306 | |||
AK140169.1 | - | 10 | 1330 | |||
AK157784.1 | - | 9 | 1316 | |||
AK166216.1 | - | 9 | 1275 |