Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026445.2 | NP_080721.1 | 3 | 356 | ||
GenBank mRNA | AK013017.1 | - | 3 | 336 | ||
AK076022.1 | - | 3 | 362 | |||
AK138300.1 | - | 3 | 347 | |||
AK140169.1 | - | 3 | 327 | |||
AK157784.1 | - | 3 | 357 | |||
AK166216.1 | - | 3 | 335 | |||
BC104404.1 | - | 1 | 465 | |||
BC104405.1 | - | 1 | 465 | |||
BC119479.1 | - | 1 | 465 | |||
BC119480.2 | - | 1 | 465 | 40050522 |