Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005859.3 | NP_001005859.1 | 2 | 144 | ||
NM_001287581.1 | NP_001274510.1 | 2 | 212 | |||
NM_026724.2 | NP_081000.1 | 2 | 280 | |||
GenBank mRNA | AK003177.1 | - | 2 | 81 | ||
AK146252.1 | - | 2 | 88 | |||
BC028517.1 | - | 1 | 54 | 1496108 | ||
BC058118.1 | - | 2 | 251 | 5683990 | ||
BC086936.1 | - | 2 | 87 | |||
CJ041925.1 | - | 2 | 88 | |||
CJ044447.1 | - | 2 | 197 |
Gene Symbol: Gm4432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG066104.2 | - | 1 | 299 |
Gene Symbol: Rpl34-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199350.1 | NP_001186279.1 | 1 | 75 | ||
GenBank mRNA | BC082285.1 | - | 1 | 70 | ||
BC100360.1 | - | 1 | 75 | 30847080 |