Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006510.4 | NP_006501.1 | 1, 2 | 778 | ||
GenBank mRNA | AB209885.1 | - | 1, 2 | 700 | ||
AF230393.1 | - | 1, 2 | 653 | |||
AF230394.1 | - | 1, 2 | 653 | |||
AJ420519.1 | - | 1, 2 | 718 | |||
AX776009.1 | - | 1, 2 | 777 | |||
AX776011.1 | - | 1, 2 | 777 | |||
BC013580.1 | - | 1, 2 | 701 | 3906095 | ||
BC066924.1 | - | 1, 2 | 749 | 4827419 | ||
J03407.1 | - | 1, 2 | 653 |