Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027187.3 | NP_081463.1 | 6 | 600 | ||
GenBank mRNA | AK010292.1 | - | 6 | 590 | ||
AK146518.1 | - | 6 | 601 | |||
AK220001.1 | - | 2 | 99 | |||
BC029252.1 | - | 6 | 630 | 3666709 | ||
BC038158.1 | - | 6 | 674 | 1493985 |